Product Details

SNP ID
rs149170140
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:152350919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAGTTTCCAGTCTCCCATGAAC[G/T]GTGGATCCTGGCTGTCTTTGTTGAG
Phenotype
MIM: 616284
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FLG-AS1 PubMed Links

Gene Details

Gene
FLG-AS1
Gene Name
FLG antisense RNA 1
There are no transcripts associated with this gene.

Gene
FLG2
Gene Name
filaggrin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014342.2 6506 Silent Mutation ACA,ACC T2289T NP_001014364.1
XM_011509531.2 6506 Silent Mutation ACA,ACC T2137T XP_011507833.1

View Full Product Details