Product Details

SNP ID
rs149210916
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:115049456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTTAATCTATGAAGAATTCTC[C/T]AGGTGGCTATACAAGAAAAATACAA
Phenotype
MIM: 613133
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN2 PubMed Links

Gene Details

Gene
TSPAN2
Gene Name
tetraspanin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308315.1 1693 UTR 3 NP_001295244.1
NM_001308316.1 1693 UTR 3 NP_001295245.1
NM_005725.5 1693 UTR 3 NP_005716.2
XM_016999996.1 1693 UTR 3 XP_016855485.1

View Full Product Details