Product Details

SNP ID
rs149354994
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:159871910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGCATTGCCTGGAATTGAAAG[A/G]GTTCTGGTGGGAGAGAGCTGAGACA
Phenotype
MIM: 605152
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP45 PubMed Links

Gene Details

Gene
CFAP45
Gene Name
cilia and flagella associated protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012337.2 Intron NP_036469.2
Gene
VSIG8
Gene Name
V-set and immunoglobulin domain containing 8
There are no transcripts associated with this gene.

View Full Product Details