Product Details
- SNP ID
-
hCV166918152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145872081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGCTCGGAGCGCCCCCGCATCTC[G/A]CTGGCCTGGCGCTCTTGTTCGGCTG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD35
PubMed Links
Gene Details
- Gene
- ANKRD35
- Gene Name
- ankyrin repeat domain 35
There are no transcripts associated with this gene.
View Full Product Details