Product Details

SNP ID
rs149458367
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:226152432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAAAATACACCCCAAACCCAAT[A/G]TCATAATTGTCTGTGGCAAATTCCC
Phenotype
MIM: 606809
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACBD3 PubMed Links

Gene Details

Gene
ACBD3
Gene Name
acyl-CoA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022735.3 1325 Silent Mutation GAC,GAT D426D NP_073572.2

View Full Product Details