Product Details

SNP ID
rs149616574
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:11938269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCCAGGATCCCTTTGCTAAGT[A/C]AGAAATCATCAACTCCATTTTGGAG
Phenotype
MIM: 153454
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLOD1 PubMed Links

Gene Details

Gene
PLOD1
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000302.3 Intron NP_000293.2
NM_001316320.1 Intron NP_001303249.1

View Full Product Details