Product Details

SNP ID
rs149627465
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:77697424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACTTCAGGCCCTCCACCATAAA[C/T]AGACTGCAGAAAATTCCATGTTTCT
Phenotype
MIM: 615146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP33 PubMed Links

Gene Details

Gene
USP33
Gene Name
ubiquitin specific peptidase 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015017.4 3096 Missense Mutation ATT,GTT I908V NP_055832.3
NM_201624.2 3096 Missense Mutation ATT,GTT I877V NP_963918.1
NM_201626.2 3096 Intron NP_963920.1
XM_005270648.1 3096 Missense Mutation ATT,GTT I900V XP_005270705.1
XM_005270649.1 3096 UTR 3 XP_005270706.1
XM_011541055.1 3096 Missense Mutation ATT,GTT I809V XP_011539357.1
XM_011541056.2 3096 Intron XP_011539358.1
XM_017000722.1 3096 Missense Mutation ATT,GTT I869V XP_016856211.1
XM_017000723.1 3096 UTR 3 XP_016856212.1
XM_017000724.1 3096 UTR 3 XP_016856213.1
XM_017000725.1 3096 Missense Mutation ATT,GTT I778V XP_016856214.1
XM_017000726.1 3096 Missense Mutation ATT,GTT I770V XP_016856215.1
XM_017000727.1 3096 Missense Mutation ATT,GTT I729V XP_016856216.1
XM_017000728.1 3096 Missense Mutation ATT,GTT I729V XP_016856217.1
XM_017000729.1 3096 UTR 3 XP_016856218.1

View Full Product Details