Product Details

SNP ID
rs149705388
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:27412656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAGTCTGACTGTGGTGGTGGC[A/G]GATAGCTACTTGCATCCACGTTTTC
Phenotype
MIM: 605875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WASF2 PubMed Links

Gene Details

Gene
WASF2
Gene Name
WAS protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201404.2 Intron NP_001188333.1
NM_006990.4 Intron NP_008921.1

View Full Product Details