Product Details

SNP ID
rs149823633
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:115286351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTTGCCCTTGATGTCTGTGG[C/T]GGTGGTCTTATCCCCAACCCACACG
Phenotype
MIM: 162030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGF PubMed Links

Gene Details

Gene
NGF
Gene Name
nerve growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002506.2 614 Missense Mutation ACC,GCC T149A NP_002497.2
XM_006710663.3 614 Missense Mutation ACC,GCC T149A XP_006710726.1
XM_011541518.2 614 Missense Mutation ACC,GCC T204A XP_011539820.1

View Full Product Details