Product Details

SNP ID
rs149829714
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:46635896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGAGGTTAAAGGTCTCCACTAA[A/C]CCGTAGGTCTCTTTCCGATCAGTAG
Phenotype
MIM: 608917
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042546.2 924 Silent Mutation GGG,GGT G244G NP_001036011.2
NM_001243728.1 924 Silent Mutation GGG,GGT G201G NP_001230657.1
NM_001256418.1 924 Silent Mutation GGG,GGT G138G NP_001243347.1
NM_022745.4 924 Silent Mutation GGG,GGT G312G NP_073582.3

View Full Product Details