Product Details

SNP ID
rs149969876
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:109737457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAAGGTGCAGGAAACGTCAC[C/T]TCAAAACGGCACATGAAAGCCTTCA
Phenotype
MIM: 138390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 889 Silent Mutation GAA,GAG E193E NP_000840.2

View Full Product Details