Product Details

SNP ID
rs150005293
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:154550040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATACATTTTTTTCATACACATCC[A/G]TCATACACTGTAACCAAAAAAAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD10 PubMed Links

Gene Details

Gene
TDRD10
Gene Name
tudor domain containing 10
There are no transcripts associated with this gene.

Gene
UBE2Q1
Gene Name
ubiquitin conjugating enzyme E2 Q1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017582.6 1758 UTR 3 NP_060052.3
Gene
UBE2Q1-AS1
Gene Name
UBE2Q1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details