Product Details

SNP ID
rs150012791
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:244411355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATTAACAGGTAGTTCTTTAAAC[A/G]CCCTTGCATTTGATATGTCTGTGTT
Phenotype
MIM: 103060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 2239 Missense Mutation GCG,GTG A417V NP_001117.2
XM_011544108.2 2239 Missense Mutation GCG,GTG A357V XP_011542410.1

View Full Product Details