Product Details

SNP ID
rs150119529
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:156251470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCCCACCTCTTTCTGGCAGC[A/G]AATGTACCTGCAGAGGAGATGTGCG
Phenotype
MIM: 112260 MIM: 614579 MIM: 610962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323614.1 2856 Missense Mutation CGC,TGC R868C NP_001310543.1
NM_001323615.1 2856 Missense Mutation CGC,TGC R875C NP_001310544.1
NM_001323616.1 2856 Missense Mutation CGC,TGC R855C NP_001310545.1
NM_001323617.1 2856 Missense Mutation CGC,TGC R855C NP_001310546.1
NM_015327.2 2856 Missense Mutation CGC,TGC R921C NP_056142.2
XM_017000842.1 2856 Missense Mutation CGC,TGC R709C XP_016856331.1
XM_017000843.1 2856 Missense Mutation CGC,TGC R660C XP_016856332.1

View Full Product Details