Product Details

SNP ID
rs150136008
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:161984110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCGCCAGGGGGTGGCCTCCTC[A/G]TAGGCCACGTCATGCAGCATGGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 2859 Silent Mutation TAC,TAT Y607Y NP_001284642.1
NM_015441.2 2859 Silent Mutation TAC,TAT Y606Y NP_056256.1
XM_005245075.2 2859 Silent Mutation TAC,TAT Y608Y XP_005245132.1
XM_011509398.2 2859 Silent Mutation TAC,TAT Y366Y XP_011507700.1
XM_017000967.1 2859 Intron XP_016856456.1

View Full Product Details