Product Details

SNP ID
rs150136942
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:115286512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCCTGAGTGTCTGCAGCTTCA[C/T]GGGGAGGCTGGGTGCTAAACAGCAC
Phenotype
MIM: 162030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGF PubMed Links

Gene Details

Gene
NGF
Gene Name
nerve growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002506.2 453 Missense Mutation CAT,CGT H95R NP_002497.2
XM_006710663.3 453 Missense Mutation CAT,CGT H95R XP_006710726.1
XM_011541518.2 453 Missense Mutation CAT,CGT H150R XP_011539820.1

View Full Product Details