Product Details

SNP ID
hCV166971377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:149887563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTTGTGACTCTCCGTTTTCTT[A/G]GGCAACAGGACAGCCTGGATATTGG
Phenotype
MIM: 615014 MIM: 602797 MIM: 601831
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST2H2AB PubMed Links

Gene Details

Gene
HIST2H2AB
Gene Name
histone cluster 2, H2ab
There are no transcripts associated with this gene.

Gene
HIST2H2AC
Gene Name
histone cluster 2, H2ac
There are no transcripts associated with this gene.

Gene
HIST2H2BE
Gene Name
histone cluster 2, H2be
There are no transcripts associated with this gene.

View Full Product Details