Product Details

SNP ID
rs150185683
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:212624918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCCGTATTATACGGCCCAGA[A/G]TGGCTCCAGCATGAGCATGTTCAAC
Phenotype
MIM: 603148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF3 PubMed Links

Gene Details

Gene
ATF3
Gene Name
activating transcription factor 3
There are no transcripts associated with this gene.

Gene
FAM71A
Gene Name
family with sequence similarity 71 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153606.3 472 Missense Mutation AAT,AGT N14S NP_705834.2

View Full Product Details