Product Details
- SNP ID
-
rs150413564
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:152409978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTCCCTGTTCTCTAGCCCCTCC[A/G]TGGCTTACAGTTTGGCTTCTGTCCT
- Phenotype
-
MIM: 611312
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRNN
PubMed Links
Gene Details
- Gene
- CRNN
- Gene Name
- cornulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016190.2 |
1178 |
Silent Mutation |
CAC,CAT |
H368H |
NP_057274.1 |
View Full Product Details