Product Details

SNP ID
rs150437510
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:161983834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTTGGTGTGGGTGTCGAAAGC[A/G]TAGGAGATGTTGGCATTCCGCTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 3135 Silent Mutation TAC,TAT Y699Y NP_001284642.1
NM_015441.2 3135 Silent Mutation TAC,TAT Y698Y NP_056256.1
XM_005245075.2 3135 Silent Mutation TAC,TAT Y700Y XP_005245132.1
XM_011509398.2 3135 Silent Mutation TAC,TAT Y458Y XP_011507700.1
XM_017000967.1 3135 Intron XP_016856456.1

View Full Product Details