Product Details

SNP ID
rs150490395
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:114720635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGGACACACAATTGGAACTT[A/G]ACGCTCTCCCCTTTCTGCAGGCAAT
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007553.2 2082 Silent Mutation GTC,GTT V652V NP_001007554.1
NM_001130523.2 2082 Silent Mutation GTC,GTT V667V NP_001123995.1
NM_001242891.1 2082 Silent Mutation GTC,GTT V698V NP_001229820.1
NM_001242892.1 2082 Silent Mutation GTC,GTT V652V NP_001229821.1
NM_001242893.1 2082 Silent Mutation GTC,GTT V621V NP_001229822.1
NM_007158.5 2082 Silent Mutation GTC,GTT V621V NP_009089.4
Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
There are no transcripts associated with this gene.

View Full Product Details