Product Details

SNP ID
rs150529054
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:152350611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTCAGAACTAGAAAATAACTGT[C/G]AATGTCTAGACAGTTGCTTGTTTGC
Phenotype
MIM: 616284
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLG-AS1 PubMed Links

Gene Details

Gene
FLG-AS1
Gene Name
FLG antisense RNA 1
There are no transcripts associated with this gene.

Gene
FLG2
Gene Name
filaggrin family member 2
There are no transcripts associated with this gene.

View Full Product Details