Product Details

SNP ID
rs150571875
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:247841456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCTGACCCCTGTGCACCAGGA[G/T]ACCACCACCAACCTGGCACAGAGCC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR11L1 PubMed Links

Gene Details

Gene
OR11L1
Gene Name
olfactory receptor family 11 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001959.1 441 Silent Mutation GTA,GTC V147V NP_001001959.1

View Full Product Details