Product Details
- SNP ID
-
rs150838067
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:77949195 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCTGTGGCATTCCCTGGGGACTT[C/G]TCTGGGCATAATAGGCTGCTTGTTG
- Phenotype
-
MIM: 603444
MIM: 613121
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FUBP1
PubMed Links
Gene Details
- Gene
- FUBP1
- Gene Name
- far upstream element binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001303433.1 |
2052 |
Missense Mutation |
ACA,AGA |
T650R |
NP_001290362.1 |
NM_003902.4 |
2052 |
Missense Mutation |
ACA,AGA |
T629R |
NP_003893.2 |
XM_011542391.2 |
2052 |
Missense Mutation |
ACA,AGA |
T650R |
XP_011540693.1 |
XM_011542392.2 |
2052 |
Missense Mutation |
ACA,AGA |
T649R |
XP_011540694.1 |
XM_011542393.2 |
2052 |
Missense Mutation |
ACA,AGA |
T629R |
XP_011540695.1 |
XM_017002739.1 |
2052 |
Missense Mutation |
ACA,AGA |
T650R |
XP_016858228.1 |
XM_017002740.1 |
2052 |
Missense Mutation |
ACA,AGA |
T650R |
XP_016858229.1 |
XM_017002741.1 |
2052 |
Missense Mutation |
ACA,AGA |
T628R |
XP_016858230.1 |
XM_017002742.1 |
2052 |
Missense Mutation |
ACA,AGA |
T649R |
XP_016858231.1 |
XM_017002743.1 |
2052 |
Missense Mutation |
ACA,AGA |
T628R |
XP_016858232.1 |
XM_017002744.1 |
2052 |
Missense Mutation |
ACA,AGA |
T628R |
XP_016858233.1 |
- Gene
- NEXN
- Gene Name
- nexilin F-actin binding protein
View Full Product Details