Product Details

SNP ID
rs151159966
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:27891843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAACATTTTAAGAAGAGAAACT[A/C]CTGAGCACTATGTAAGTACTCTCCT
Phenotype
MIM: 179836
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPA2 PubMed Links

Gene Details

Gene
RPA2
Gene Name
replication protein A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286076.1 1487 UTR 3 NP_001273005.1
NM_001297558.1 1487 Intron NP_001284487.1
NM_002946.4 1487 UTR 3 NP_002937.1
Gene
THEMIS2
Gene Name
thymocyte selection associated family member 2
There are no transcripts associated with this gene.

View Full Product Details