Product Details

SNP ID
rs137949815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33083530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGATATGGTGAGATCCTTGAGTC[A/C]GAGGGAAGCATCAGGGACCTCCGAA
Phenotype
MIM: 615717 MIM: 615718
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BPIFB3 PubMed Links

Gene Details

Gene
BPIFB3
Gene Name
BPI fold containing family B member 3
There are no transcripts associated with this gene.

Gene
BPIFB4
Gene Name
BPI fold containing family B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182519.2 348 Silent Mutation TCA,TCC S111S NP_872325.2

View Full Product Details