Product Details

SNP ID
rs138720031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46062809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTAGCAGAGGGGAGCACTTGGC[C/T]GCTCTGGAGCGGTTGGGAGCTTGGC
Phenotype
MIM: 616825 MIM: 606726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCOA5 PubMed Links

Gene Details

Gene
NCOA5
Gene Name
nuclear receptor coactivator 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020967.2 1362 Missense Mutation NP_066018.1
XM_005260474.3 1362 Missense Mutation XP_005260531.1
XM_011528951.2 1362 UTR 3 XP_011527253.1
XM_017027988.1 1362 UTR 3 XP_016883477.1
Gene
SLC12A5
Gene Name
solute carrier family 12 member 5
There are no transcripts associated with this gene.

View Full Product Details