Product Details
- SNP ID
-
rs139093108
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63747066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGTCGTCCTCGCCCAGCTCCTCC[A/G]CCTCCCCTCGTGGGTCCTCAGGGTC
- Phenotype
-
MIM: 609809
MIM: 609493
MIM: 614639
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LIME1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74552928] are located under a probe and SNP(s) [rs12625387] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LIME1
- Gene Name
- Lck interacting transmembrane adaptor 1
There are no transcripts associated with this gene.
- Gene
- SLC2A4RG
- Gene Name
- SLC2A4 regulator
There are no transcripts associated with this gene.
- Gene
- ZBTB46
- Gene Name
- zinc finger and BTB domain containing 46
View Full Product Details