Product Details

SNP ID
rs139294924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56004256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCGGGATCCCGGTGCTCGGGAA[A/G]ATGCTAGCGGCTAGGTCGACAGCGC
Phenotype
MIM: 615029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLN4 PubMed Links

Gene Details

Gene
CBLN4
Gene Name
cerebellin 4 precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080617.5 1217 UTR 5 NP_542184.1

View Full Product Details