Product Details

SNP ID
rs139459928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64169208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGTGGGAGTTTGGATGCTCC[A/C]TGGTGCACCAAGGCCTGCTTTAGAA
Phenotype
MIM: 600379
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYT1 PubMed Links

Gene Details

Gene
MYT1
Gene Name
myelin transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004535.2 Intron NP_004526.1

View Full Product Details