Product Details

SNP ID
rs140118504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63696983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAGCCCCACGACGTGTGGCCC[A/G]TGTCCACCGCGCCACTACACGCAGT
Phenotype
MIM: 604699 MIM: 608833 MIM: 603361
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFRP1 PubMed Links

Gene Details

Gene
ARFRP1
Gene Name
ADP ribosylation factor related protein 1
There are no transcripts associated with this gene.

Gene
RTEL1
Gene Name
regulator of telomere elongation helicase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283009.1 333 Intron NP_001269938.1
NM_001283010.1 333 Intron NP_001269939.1
NM_016434.3 333 Intron NP_057518.1
NM_032957.4 333 Intron NP_116575.3
Gene
RTEL1-TNFRSF6B
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNFRSF6B
Gene Name
TNF receptor superfamily member 6b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003823.3 333 Silent Mutation CCA,CCG P72P NP_003814.1

View Full Product Details