Product Details

SNP ID
rs140312685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18294320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATACCCTTCAAAAGCGCCAAGTT[C/T]GTGAAAGATGAGAAAAGACAGAGGA
Phenotype
MIM: 604075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 4503 Intron NP_001076799.1
NM_001282995.1 4503 Intron NP_001269924.1
NM_001282996.1 4503 Intron NP_001269925.1
NM_001282997.1 4503 Intron NP_001269926.1
NM_001282998.1 4503 Intron NP_001269927.1
NM_001282999.1 4503 Intron NP_001269928.1
NM_001283000.1 4503 Intron NP_001269929.1
NM_001283001.1 4503 Intron NP_001269930.1
NM_001283002.1 4503 Intron NP_001269931.1
NM_001283003.1 4503 Intron NP_001269932.1
NM_001283004.1 4503 Intron NP_001269933.1
NM_001283005.1 4503 Intron NP_001269934.1
NM_001283006.1 4503 Intron NP_001269935.1
NM_001283007.1 4503 Intron NP_001269936.1
NM_001283008.1 4503 Intron NP_001269937.1
NM_003434.5 4503 Intron NP_003425.2
XM_005260819.1 4503 Intron XP_005260876.1
XM_005260820.4 4503 Intron XP_005260877.1
XM_011529336.1 4503 Intron XP_011527638.1
XM_011529337.1 4503 Intron XP_011527639.1
XM_011529338.2 4503 Intron XP_011527640.1
XM_011529339.1 4503 Intron XP_011527641.1
XM_011529340.1 4503 Intron XP_011527642.1
XM_017028041.1 4503 UTR 5 XP_016883530.1
XM_017028042.1 4503 Intron XP_016883531.1
XM_017028043.1 4503 UTR 5 XP_016883532.1
XM_017028044.1 4503 UTR 5 XP_016883533.1
XM_017028045.1 4503 UTR 5 XP_016883534.1
XM_017028046.1 4503 UTR 5 XP_016883535.1
XM_017028047.1 4503 Intron XP_016883536.1
XM_017028048.1 4503 Intron XP_016883537.1
XM_017028049.1 4503 UTR 5 XP_016883538.1
XM_017028050.1 4503 Intron XP_016883539.1
XM_017028051.1 4503 Intron XP_016883540.1
XM_017028052.1 4503 Intron XP_016883541.1
XM_017028053.1 4503 Intron XP_016883542.1
XM_017028054.1 4503 Intron XP_016883543.1
XM_017028055.1 4503 Intron XP_016883544.1
XM_017028056.1 4503 Intron XP_016883545.1
XM_017028057.1 4503 Intron XP_016883546.1
XM_017028058.1 4503 Intron XP_016883547.1

View Full Product Details