Product Details

SNP ID
rs140971821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:7982374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCCTCAGGCTCTACTTCCTC[C/T]CGGGTCACACCGTCCTCTCCTGGGG
Phenotype
MIM: 616766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMX4 PubMed Links

Gene Details

Gene
TMX4
Gene Name
thioredoxin related transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021156.3 1143 Silent Mutation CGA,CGG R309R NP_066979.2

View Full Product Details