Product Details
- SNP ID
-
rs142807270
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46009055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGAACCAATCTCACCGACAGGCA[G/T]CTGGCAGAGGTGGGCAAACACCTAG
- Phenotype
-
MIM: 120361
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MMP9
PubMed Links
Gene Details
- Gene
- MMP9
- Gene Name
- matrix metallopeptidase 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004994.2 |
148 |
Missense Mutation |
CAG,CAT |
Q43H |
NP_004985.2 |
View Full Product Details