Product Details
- SNP ID
-
rs145523811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:3690137 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCACCCTGGGGCTGACTGGAGGC[C/T]ACCAGTCGACTGCCAAGGTGGAGAG
- Phenotype
-
MIM: 607114
MIM: 600751
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM33
PubMed Links
Gene Details
- Gene
- ADAM33
- Gene Name
- ADAM metallopeptidase domain 33
There are no transcripts associated with this gene.
- Gene
- SIGLEC1
- Gene Name
- sialic acid binding Ig like lectin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023068.3 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
NP_075556.1 |
XM_006723610.3 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_006723673.1 |
XM_011529324.2 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527626.1 |
XM_011529325.2 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527627.1 |
XM_011529326.2 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527628.1 |
XM_011529327.2 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527629.1 |
XM_011529328.2 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527630.1 |
XM_011529329.1 |
6100 |
Silent Mutation |
GTA,GTG |
V1573V |
XP_011527631.1 |
XM_011529330.1 |
6100 |
Silent Mutation |
GTA,GTG |
V1487V |
XP_011527632.1 |
XM_011529331.1 |
6100 |
Silent Mutation |
GTA,GTG |
V1484V |
XP_011527633.1 |
XM_011529332.1 |
6100 |
Silent Mutation |
GTA,GTG |
V1370V |
XP_011527634.1 |
XM_011529333.2 |
6100 |
Intron |
|
|
XP_011527635.1 |
View Full Product Details