Product Details

SNP ID
rs145944666
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35286912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGTCTGCCGCCGCTCATCTG[A/G]TGCCCGTCTGGCAGGTCTGGCTGAG
Phenotype
MIM: 602912 MIM: 604871
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EIF6 PubMed Links
Additional Information
For this assay, SNP(s) [rs2425049] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EIF6
Gene Name
eukaryotic translation initiation factor 6
There are no transcripts associated with this gene.

Gene
FAM83C
Gene Name
family with sequence similarity 83 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178468.5 1985 Missense Mutation CCA,TCA P623S NP_848563.1
Gene
FAM83C-AS1
Gene Name
FAM83C antisense RNA 1
There are no transcripts associated with this gene.

Gene
MMP24
Gene Name
matrix metallopeptidase 24
There are no transcripts associated with this gene.

Gene
MMP24-AS1
Gene Name
MMP24 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details