Product Details

SNP ID
rs147555737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45709273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAATTTTTCATGCTTAAAATA[A/G]AGATAACACCACTTACCTTACGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3617 PubMed Links

Gene Details

Gene
MIR3617
Gene Name
microRNA 3617
There are no transcripts associated with this gene.

Gene
WFDC10B
Gene Name
WAP four-disulfide core domain 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172006.2 Intron NP_742003.1
NM_172131.2 Intron NP_742143.1
XM_017027824.1 Intron XP_016883313.1
XM_017027825.1 Intron XP_016883314.1
Gene
WFDC13
Gene Name
WAP four-disulfide core domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172005.1 Intron NP_742002.1
Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 Intron NP_542181.1
XM_011528553.2 Intron XP_011526855.1
XM_017027668.1 Intron XP_016883157.1

View Full Product Details