Product Details

SNP ID
rs147688404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10405271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGTTTTTATCTTCAATAACATA[A/T]GAAAGATCCAAAATCAAATTGGCTG
Phenotype
MIM: 604896
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MKKS PubMed Links

Gene Details

Gene
MKKS
Gene Name
McKusick-Kaufman syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018848.3 2579 Silent Mutation TCA,TCT S563S NP_061336.1
NM_170784.2 2579 Silent Mutation TCA,TCT S563S NP_740754.1

View Full Product Details