Product Details

SNP ID
rs147913483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18487611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAATCTTAATTATTTTTGATT[A/G]TAAAATACATGCTTAGTGTAACAAA
Phenotype
MIM: 602908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR3F PubMed Links

Gene Details

Gene
POLR3F
Gene Name
RNA polymerase III subunit F
There are no transcripts associated with this gene.

Gene
RBBP9
Gene Name
RB binding protein 9, serine hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006606.2 2790 UTR 3 NP_006597.2
XM_005260652.2 2790 UTR 3 XP_005260709.1

View Full Product Details