Product Details

SNP ID
rs148109555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64037425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGAGTTGAATGGGCATGGCCAT[C/G]AGTGAGACAGGCTGTACTGGGACAA
Phenotype
MIM: 613979 MIM: 604784
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00176 PubMed Links

Gene Details

Gene
LINC00176
Gene Name
long intergenic non-protein coding RNA 176
There are no transcripts associated with this gene.

Gene
PRPF6
Gene Name
pre-mRNA processing factor 6
There are no transcripts associated with this gene.

Gene
TCEA2
Gene Name
transcription elongation factor A2
There are no transcripts associated with this gene.

View Full Product Details