Product Details

SNP ID
rs149095209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10405321 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTACAGCCACCTGTAGGCCAC[C/T]AAGCTTTGCAGTCAAACAGTCCAAG
Phenotype
MIM: 604896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKKS PubMed Links

Gene Details

Gene
MKKS
Gene Name
McKusick-Kaufman syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018848.3 2529 Missense Mutation NP_061336.1
NM_170784.2 2529 Missense Mutation NP_740754.1

View Full Product Details