Product Details

SNP ID
rs149230426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36609681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGATGAGGAAGAATATGGTGGCA[C/T]GTGGGAGCTCACGAAGGAAGGGTTT
Phenotype
MIM: 606577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf24 PubMed Links

Gene Details

Gene
C20orf24
Gene Name
chromosome 20 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199534.1 493 Missense Mutation ACG,ATG T100M NP_001186463.1
NM_018840.4 493 Missense Mutation ACG,ATG T100M NP_061328.1
NM_199483.2 493 Intron NP_955777.1
Gene
SLA2
Gene Name
Src like adaptor 2
There are no transcripts associated with this gene.

Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199535.1 493 Missense Mutation ACG,ATG T126M NP_001186464.1

View Full Product Details