Product Details

SNP ID
rs149887460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59939163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCACCCGCCACTTGGTAGCGCA[C/T]CGCGAAGTGCACGCGGGAGCCGAGC
Phenotype
MIM: 603326 MIM: 604105
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM217B PubMed Links

Gene Details

Gene
FAM217B
Gene Name
family with sequence similarity 217 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190826.1 1135 Intron NP_001177755.1
NM_001190827.1 1135 Intron NP_001177756.1
NM_022106.2 1135 Intron NP_071389.1
XM_011528985.2 1135 Intron XP_011527287.1
XM_011528986.2 1135 Intron XP_011527288.1
Gene
PPP1R3D
Gene Name
protein phosphatase 1 regulatory subunit 3D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006242.3 1135 Missense Mutation ATG,GTG M257V NP_006233.1
Gene
SYCP2
Gene Name
synaptonemal complex protein 2
There are no transcripts associated with this gene.

View Full Product Details