Product Details

SNP ID
rs151272379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63277226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTGGTCGCTCTGGCCGAAGGC[A/G]GAGAGTGGTCTCTGGAGTCATCACC
Phenotype
MIM: 608377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP1 PubMed Links

Gene Details

Gene
ARFGAP1
Gene Name
ADP ribosylation factor GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281482.1 570 Missense Mutation AGA,GGA R122G NP_001268411.1
NM_001281483.1 570 Missense Mutation AGA,GGA R69G NP_001268412.1
NM_001281484.1 570 Intron NP_001268413.1
NM_018209.3 570 Missense Mutation AGA,GGA R122G NP_060679.1
NM_175609.2 570 Missense Mutation AGA,GGA R122G NP_783202.1
XM_006723823.2 570 Missense Mutation AGA,GGA R122G XP_006723886.1
XM_011528901.1 570 Missense Mutation AGA,GGA R122G XP_011527203.1
XM_017027950.1 570 Missense Mutation AGA,GGA R122G XP_016883439.1
Gene
MIR4326
Gene Name
microRNA 4326
There are no transcripts associated with this gene.

View Full Product Details