Product Details
- SNP ID
-
rs138685842
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42363272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCATACCTTCTGGAGGGACGTCG[A/T]TGGTATTAGGATAGAAGCACCAGGG
- Phenotype
-
MIM: 113710
MIM: 605511
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TFF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61735845] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFF1
- Gene Name
- trefoil factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003225.2 |
261 |
Missense Mutation |
AAC,ATC |
N74I |
NP_003216.1 |
- Gene
- TMPRSS3
- Gene Name
- transmembrane protease, serine 3
There are no transcripts associated with this gene.
View Full Product Details