Product Details
- SNP ID
-
rs138939112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:33907373 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGTCACTGTGCAAGGTACCTCTC[C/T]GCGATGGACACTCATCATGGTAGAA
- Phenotype
-
MIM: 600828
MIM: 602442
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP5O
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62226670] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP5O
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001697.2 |
|
Intron |
|
|
NP_001688.1 |
- Gene
- ITSN1
- Gene Name
- intersectin 1
There are no transcripts associated with this gene.
View Full Product Details