Product Details

SNP ID
rs141518595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43669932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAATCTGGAAGGGGCTCTTCT[A/C]CTGCATGTGGGTGCAGGATGAACCC
Phenotype
MIM: 610654
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RRP1B PubMed Links

Gene Details

Gene
RRP1B
Gene Name
ribosomal RNA processing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015056.2 293 Missense Mutation TAC,TCC Y60S NP_055871.1

View Full Product Details