Product Details

SNP ID
rs141793094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30396603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGTGGATCAGGCTTCTACTAT[C/T]GATCATCTTGTTAAATTGCTGATTT
Phenotype
MIM: 608718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-1 PubMed Links

Gene Details

Gene
KRTAP13-1
Gene Name
keratin associated protein 13-1
There are no transcripts associated with this gene.

View Full Product Details