Product Details

SNP ID
rs141942958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14219647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTGTCTGCTTTAAACACCCA[A/G]AATCGGTGTCTTATGCCATAGCTTT
Phenotype
MIM: 609252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPI PubMed Links

Gene Details

Gene
LIPI
Gene Name
lipase I
There are no transcripts associated with this gene.

Gene
RBM11
Gene Name
RNA binding motif protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320602.1 400 Missense Mutation AAA,GAA K61E NP_001307531.1
NM_144770.4 400 Missense Mutation AAA,GAA K61E NP_658983.3
XM_005260997.4 400 Missense Mutation AAA,GAA K61E XP_005261054.1
XM_017028384.1 400 Intron XP_016883873.1
XM_017028385.1 400 UTR 5 XP_016883874.1
XM_017028386.1 400 UTR 5 XP_016883875.1
XM_017028387.1 400 UTR 5 XP_016883876.1

View Full Product Details