Product Details
- SNP ID
-
rs142678238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:30440437 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTACCCCAGCAATGCCATCTATT[A/C]TCCAAATACCTGCCAACTGGGCTCC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP13-4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2832873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP13-4
- Gene Name
- keratin associated protein 13-4
There are no transcripts associated with this gene.
- Gene
- KRTAP15-1
- Gene Name
- keratin associated protein 15-1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181623.1 |
110 |
Missense Mutation |
TAT,TCT |
Y37S |
NP_853654.1 |
View Full Product Details